1-(Difluoromethyl)-2,4-difluorobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Difluoro-1-(difluoromethyl)benzene

035674-1G 035674-5G
101839-632EA 38.46 USD
101839-632 101839-634
1-(Difluoromethyl)-2,4-difluorobenzene ≥97%
1-(Difluoromethyl)-2,4-difluorobenzene

Hazard: Irritant

Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 66…68 °C (100 Torr)
MDL Number: MFCD06657968
CAS Number: 845866-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR