4-Bromo-1-(difluoromethyl)-2-fluorobenzene ≥98%

Supplier: Matrix Scientific

035672-1G 035672-5G
101839-624EA 67.69 USD
101839-624 101839-626
4-Bromo-1-(difluoromethyl)-2-fluorobenzene ≥98%
4-Bromo-1-(difluoromethyl)-2-fluorobenzene

Hazard: Irritant

Formula: C₇H₄BrF₃
MDL Number: MFCD06657963
CAS Number: 749932-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR