2-(2-Pyrrolidinyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Pyrrolidin-2-ylpyridine

035664-250MG 035664-1G
101839-602EA 1320 USD
101839-602 101839-604
2-(2-Pyrrolidinyl)pyridine ≥95%
2-(2-Pyrrolidinyl)pyridine

Hazard: Irritant/Store in Freezer

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
MDL Number: MFCD01691538
CAS Number: 77790-61-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR