5-Methyl-3-pyridineboronic acid

Supplier: Matrix Scientific

Synonyms: 5-Methylpyridine-3-boronic acid

035646-1G 035646-5G
101839-588EA 67.69 USD
101839-588 101839-590
5-Methyl-3-pyridineboronic acid
5-Methyl-3-pyridineboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₆H₈BNO₂
MW: 136.95 g/mol
MDL Number: MFCD03428528
CAS Number: 173999-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR