4-Pentylphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-n-Pentylbenzeneboronic acid, (4-Pentylphenyl)boronic acid

035619-1G 035619-5G 035619-25G
101839-574EA 15.38 USD
101839-574 101839-576 101839-578
4-Pentylphenylboronic acid ≥95%
4-Pentylphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₇BO₂
MW: 192.07 g/mol
MDL Number: MFCD00995151
CAS Number: 121219-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR