4-Isopropylphenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Isopropylbenzeneboronic acid

035612-5G 035612-25G
101839-568EA 40 USD
101839-568 101839-570
4-Isopropylphenylboronic acid ≥98%
4-Isopropylphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
MDL Number: MFCD01074614
CAS Number: 16152-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR