4-Fluoro-3-methoxyphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Fluoro-3-methoxyphenyl)boronic acid, 4-Fluoro-3-methoxybenzeneboronic acid

035600-1G 035600-5G
101839-548EA 35.38 USD
101839-548 101839-550
4-Fluoro-3-methoxyphenylboronic acid ≥95%
4-Fluoro-3-methoxyphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD08056358
CAS Number: 854778-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR