4-Carboxy-2-nitrophenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-Carboxy-2-nitrobenzeneboronic acid

035566-1G 035566-5G
101839-512EA 44.62 USD
101839-512 101839-514
4-Carboxy-2-nitrophenylboronic acid
4-Carboxy-2-nitrophenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₆BNO₆
MW: 210.94 g/mol
MDL Number: MFCD00448780
CAS Number: 85107-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR