(4-Boc-aminophenyl)boronic acid
Supplier: Matrix Scientific
Synonyms:
(4-Boc-Aminophenyl)Boronic Acid, 4-Boc-aminophenylboronic acid, (4-Boc-Aminophenyl)boronic acid, 4-(N-BOC-amino)phenylboronic acid, [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic Acid
035559-1G
035559-5G
101839-506EA
20
USD
101839-506
101839-508
(4-Boc-aminophenyl)boronic acid
(4-Boc-aminophenyl)boronic acid
Hazard: Irritant/Refrigerate
Formula:
C₁₁H₁₆BNO₄ MW: 237.062 g/mol |
MDL Number:
MFCD02093054 CAS Number: 380430-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...