3-Methoxy-4-carboxyphenylboronic acid

Supplier: Matrix Scientific

035536-1G 035536-5G
101839-488EA 129.23 USD
101839-488 101839-490
3-Methoxy-4-carboxyphenylboronic acid
3-Methoxy-4-carboxyphenylboronic acid

Hazard: Irritant

Formula: C₈H₉BO₅
MDL Number: MFCD06656278
CAS Number: 851335-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR