3-Chloro-5-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Chloro-5-(trifluoromethyl)phenyl)boronic acid

035513-1G 035513-5G
101839-476EA 43.08 USD
101839-476 101839-478
3-Chloro-5-(trifluoromethyl)phenylboronic acid ≥95%
3-Chloro-5-(trifluoromethyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₅BClF₃O₂
MDL Number: MFCD07778020
CAS Number: 1160561-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR