4-Benzyloxy-3-chlorophenylboronic acid

Supplier: Matrix Scientific

Synonyms: 4-Benzyloxy-3-chlorobenzeneboronic acid, (4-(Benzyloxy)-3-chlorophenyl)boronic acid

035506-5G 035506-25G
101839-472EA 93.85 USD
101839-472 101839-474
4-Benzyloxy-3-chlorophenylboronic acid
4-Benzyloxy-3-chlorophenylboronic acid

Hazard: Irritant

Formula: C₁₃H₁₂BClO₃
MW: 262.50 g/mol
MDL Number: MFCD04115642
CAS Number: 845551-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR