3-Carboxy-4-fluorophenylboronic acid
Supplier: Matrix Scientific
Synonyms:
3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid
035502-1G
035502-5G
101839-468EA
63.08
USD
101839-468
101839-470
3-Carboxy-4-fluorophenylboronic acid
3-Carboxy-4-fluorophenylboronic acid
Hazard: Irritant/Refrigerate
Formula:
C₇H₆BFO₄ MW: 183.93 g/mol |
MDL Number:
MFCD03095129 CAS Number: 872460-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...