2-Fluoro-4-methoxyphenylboronic acid

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-methoxybenzeneboronic acid

035453-1G 035453-5G 035453-25G
101839-428EA 30.77 USD
101839-428 101839-430 101839-432
2-Fluoro-4-methoxyphenylboronic acid
2-Fluoro-4-methoxyphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD04039323
CAS Number: 162101-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR