2-Fluoro-(4-methoxycarbonyl)phenylboronic acid

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid

035452-1G 035452-5G
101839-422EA 44.62 USD
101839-422 101839-424
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Melting Pt: 173-175°C
MDL Number: MFCD08436034
CAS Number: 603122-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR