2-Fluoro-4-biphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-4-biphenylylboronic acid, (3-fluoro-4-phenyl)phenylboronic acid, 3-Fluoro-4-phenylbenzeneboronic acid, 2-Fluoro-1,1'-biphenyl-4-ylboronic acid

035446-5G 035446-25G
101839-416EA 44.62 USD
101839-416 101839-418
2-Fluoro-4-biphenylboronic acid ≥97%
2-Fluoro-4-biphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₂H₁₀BFO₂
MW: 216.02 g/mol
MDL Number: MFCD01075707
CAS Number: 178305-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR