2-Chloro-5-(hydroxymethyl)phenylboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-5-hydroxymethylphenylboronic acid
035397-1G
035397-5G
101839-402EA
138.46
USD
101839-402
101839-404
2-Chloro-5-(hydroxymethyl)phenylboronic acid ≥95%
2-Chloro-5-(hydroxymethyl)phenylboronic acid
Hazard: Irritant/Refrigerate
Formula:
C₇H₈BClO₃ MW: 186.4000 g/mol |
MDL Number:
MFCD06656270 CAS Number: 1003042-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...