2-Chloro-5-bromo-4-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-chloro-4-(trifluoromethyl)phenylboronic acid, (5-Bromo-2-chloro-4-(trifluoromethyl)phenyl)boronic acid

035393-250MG 035393-1G
101839-398EA 138.46 USD
101839-398 101839-400
2-Chloro-5-bromo-4-(trifluoromethyl)phenylboronic acid ≥95%
2-Chloro-5-bromo-4-(trifluoromethyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₄BBrClF₃O₂
MW: 303.28 g/mol
MDL Number: MFCD12026719
CAS Number: 1452574-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR