2-Chloro-4-methoxyphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-methoxybenzeneboronic acid

035388-1G 035388-5G
101839-390EA 121.54 USD
101839-390 101839-392
2-Chloro-4-methoxyphenylboronic acid ≥95%
2-Chloro-4-methoxyphenylboronic acid

Hazard: Irritant

Formula: C₇H₈BClO₃
MW: 186.40 g/mol
MDL Number: MFCD03411935
CAS Number: 219735-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR