2-Chloro-6-iodopyridin-3-amine ≥95%
Supplier: Matrix Scientific
035244-100MG
035244-500MG
101839-266EA
244.7
USD
101839-266
101839-268
2-Chloro-6-iodopyridin-3-amine ≥95%
2-Chloro-6-iodopyridin-3-amine
Hazard: Irritant
Formula:
C₅H₄ClIN₂ Boiling Pt: 340°C Density: 2.139 Flash Pt: 159°C |
MDL Number:
MFCD11044245 CAS Number: 1032507-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...