5-Chloro-2-fluoronicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-chloropyridine-3-carboxylic acid

034850-1G 034850-5G
101838-498EA 98.46 USD
101838-498 101838-500
5-Chloro-2-fluoronicotinic acid ≥98%
5-Chloro-2-fluoronicotinic acid

Hazard: Irritant

Formula: C₆H₃ClFNO₂
MDL Number: MFCD04112518
CAS Number: 884494-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR