5-Chloro-6-methoxypyridine-3-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-6-methoxynicotinic acid

034844-5G 034844-25G
101838-486EA 75.38 USD
101838-486 101838-488
5-Chloro-6-methoxypyridine-3-carboxylic acid ≥98%
5-Chloro-6-methoxypyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD04972405
CAS Number: 884494-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR