1-(Difluoromethyl)-2-fluorobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Difluoromethyl-1-fluorobenzene

034761-1G 034761-5G
101838-312EA 72.31 USD
101838-312 101838-314
1-(Difluoromethyl)-2-fluorobenzene ≥97%
1-(Difluoromethyl)-2-fluorobenzene

Hazard: Irritant

Formula: C₇H₅F₃
MW: 146.112 g/mol
MDL Number: MFCD06657985
CAS Number: 63878-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR