1-Bromo-5-(difluoromethyl)-2-fluorobenzene ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-4-(difluoromethyl)-1-fluorobenzene
034758-1G
034758-5G
101838-308EA
75.38
USD
101838-308
101838-310
1-Bromo-5-(difluoromethyl)-2-fluorobenzene ≥97%
1-Bromo-5-(difluoromethyl)-2-fluorobenzene
Hazard: Irritant
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol |
MDL Number:
MFCD08276255 CAS Number: 886509-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...