2,4,5-Trifluoropyrimidine ≥98%

Supplier: Matrix Scientific

034747-1G 034747-5G
101838-278EA 89.23 USD
101838-278 101838-280
2,4,5-Trifluoropyrimidine ≥98%
2,4,5-Trifluoropyrimidine

Hazard: Toxic

Formula: C₄HF₃N₂
MDL Number: MFCD11845791
CAS Number: 17573-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR