Ethyl-3-cyclopropyl-3-oxopropanoate ≥97%
Supplier: Matrix Scientific
034733-5G
034733-25G
101838-246EA
15.38
USD
101838-246
101838-248
Ethyl-3-cyclopropyl-3-oxopropanoate ≥97%
Ethyl-3-cyclopropyl-3-oxopropanoate
Hazard: Irritant
Formula:
C₈H₁₂O₃ Boiling Pt: 73°C/1mm |
MDL Number:
MFCD00184570 CAS Number: 24922-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...