4-Bromo-1,5-dimethyl-1H-pyrazole, Technical Grade

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1,5-dimethylpyrazole

034680-1G 034680-5G
101838-138EA 90.77 USD
101838-138 101838-140
4-Bromo-1,5-dimethyl-1H-pyrazole, Technical Grade
4-Bromo-1,5-dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₇BrN₂
MW: 175.0265 g/mol
MDL Number: MFCD09759006
CAS Number: 5775-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR