2-Methyl-4-nitrobenzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-nitro-1-(trifluoromethyl)benzene

032535-1G 032535-5G
101833-942EA 121.54 USD
101833-942 101833-944
2-Methyl-4-nitrobenzotrifluoride ≥97%
2-Methyl-4-nitrobenzotrifluoride

Hazard: Irritant-Harmful

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
MDL Number: MFCD11226539
CAS Number: 1960-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR