4-Chloro-2,6-difluorobenzaldehyde ≥97%

Supplier: Matrix Scientific

032525-1G 032525-5G
101833-922EA 52.31 USD
101833-922 101833-924
4-Chloro-2,6-difluorobenzaldehyde ≥97%
4-Chloro-2,6-difluorobenzaldehyde

Hazard: Irritant/Air Sensitive/Refrigerate

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Melting Pt: 70-74°C
MDL Number: MFCD03094460
CAS Number: 252004-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR