2-Amino-3-fluoro-5-(trifluoromethyl)pyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-5-(trifluoromethyl)pyridin-2-amine
032518-1G
032518-5G
101833-908EA
92.31
USD
101833-908
101833-910
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine ≥98%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Hazard: Toxic
Formula:
C₆H₄F₄N₂ Melting Pt: 64-68°C |
MDL Number:
MFCD10699117 CAS Number: 852062-17-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...