5-Bromo-2-methox-6-methylypyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-6-methoxy-2-picoline, 3-Bromo-6-methoxy-2-methylpyridine

032517-1G 032517-5G
101833-904EA 212.31 USD
101833-904 101833-906
5-Bromo-2-methox-6-methylypyridine ≥98%
5-Bromo-2-methox-6-methylypyridine

Hazard: Irritant

Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD03094943
CAS Number: 126717-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR