5-Aminopyrimidine ≥97%

Supplier: Matrix Scientific

032476-1G 032476-5G
101833-816EA 26.15 USD
101833-816 101833-818
5-Aminopyrimidine ≥97%
5-Aminopyrimidine

Hazard: Irritant-Harmful

Formula: C₄H₅N₃
MW: 95.1000 g/mol
MDL Number: MFCD01529870
CAS Number: 591-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR