4-(1H-Pyrazol-3-yl)benzonitrile ≥97%

Supplier: Matrix Scientific

032434-1G 032434-5G
101833-716EA 170.77 USD
101833-716 101833-718
4-(1H-Pyrazol-3-yl)benzonitrile ≥97%
4-(1H-Pyrazol-3-yl)benzonitrile

Hazard: Irritant

Formula: C₁₀H₇N₃
MDL Number: MFCD01569438
CAS Number: 474706-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR