Ethyl-2-fluoroisobutyrate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-fluoro-2-methylpropanoate

032433-1G 032433-5G
101833-712EA 38.46 USD
101833-712 101833-714
Ethyl-2-fluoroisobutyrate ≥97%
Ethyl-2-fluoroisobutyrate

Hazard: Flammable/Toxic/Refrigerate/Lachrymator

Formula: C₆H₁₁FO₂
MW: 134.15 g/mol
Boiling Pt: 120…126 °C
MDL Number: MFCD08062432
CAS Number: 55816-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR