2-Bromo-6-methylbenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-methyl-2-(trifluoromethyl)benzene

032422-1G 032422-5G
101833-690EA 153.85 USD
101833-690 101833-692
2-Bromo-6-methylbenzotrifluoride ≥95%
2-Bromo-6-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631583
CAS Number: 944268-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR