3-Chloro-2-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
032393-250MG
032393-1G
101833-630EA
66.15
USD
101833-630
101833-632
3-Chloro-2-(trifluoromethyl)pyridine ≥97%
3-Chloro-2-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₃ClF₃N Boiling Pt: 86…88 °C (40 mmHg) |
MDL Number:
MFCD11044298 CAS Number: 749875-32-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...