2-Methyl-6-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

032390-1G 032390-5G
101833-622EA 36.92 USD
101833-622 101833-624
2-Methyl-6-(trifluoromethyl)pyridine ≥97%
2-Methyl-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₇H₆F₃N
MW: 161.13 g/mol
MDL Number: MFCD07774130
CAS Number: 1620-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR