4-Bromo-2-methylbenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-methyl-1-(trifluoromethyl)benzene

032375-1G 032375-5G
101833-594EA 36.92 USD
101833-594 101833-596
4-Bromo-2-methylbenzotrifluoride ≥95%
4-Bromo-2-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₆BrF₃
MW: 239.0325 g/mol
MDL Number: MFCD11100526
CAS Number: 936092-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR