3-Fluoro-2-hydroxy-5-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-5-(trifluoromethyl)pyridin-2-ol

032350-1G 032350-5G
101833-530EA 127.69 USD
101833-530 101833-532
3-Fluoro-2-hydroxy-5-(trifluoromethyl)pyridine ≥98%
3-Fluoro-2-hydroxy-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃F₄NO
MW: 181.09 g/mol
MDL Number: MFCD10699119
CAS Number: 1040683-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR