5-Bromo-2-methoxy-3-nitropyridine

Supplier: Matrix Scientific

032316-5G 032316-25G
101833-452EA 186.15 USD
101833-452 101833-454
5-Bromo-2-methoxy-3-nitropyridine
5-Bromo-2-methoxy-3-nitropyridine

Hazard: Irritant

Formula: C₆H₅BrN₂O₃
MW: 233.0200 g/mol
MDL Number: MFCD07374968
CAS Number: 152684-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR