4,6-Dimethoxy-2-mercaptopyrimidine ≥98%

Supplier: Matrix Scientific

031267-5G 031267-25G
101831-354EA 80 USD
101831-354 101831-356
4,6-Dimethoxy-2-mercaptopyrimidine ≥98%
4,6-Dimethoxy-2-mercaptopyrimidine

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MDL Number: MFCD00672150
CAS Number: 57235-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR