4,6-Dimethyl-2-methylsulfonylpyrimidine ≥98%
Supplier: Matrix Scientific
031256-25G
031256-100G
101831-330EA
60
USD
101831-330
101831-332
4,6-Dimethyl-2-methylsulfonylpyrimidine ≥98%
4,6-Dimethyl-2-methylsulfonylpyrimidine
Hazard: Irritant
Formula:
C₇H₁₀N₂O₂S MW: 186.23 g/mol |
MDL Number:
MFCD03788286 CAS Number: 35144-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...