4-(4-Formylphenoxy)butanoic acid

Supplier: Matrix Scientific

031184-500MG 031184-2.5G
101831-280EA 461.54 USD
101831-280 101831-282
4-(4-Formylphenoxy)butanoic acid
4-(4-Formylphenoxy)butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₄
MDL Number: MFCD02258439
CAS Number: 99865-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR