1-Benzoylpiperidin-4-amine hydrochloride

Supplier: Matrix Scientific

031025-500MG 031025-1G
101831-224EA 250.77 USD
101831-224 101831-226
1-Benzoylpiperidin-4-amine hydrochloride
1-Benzoylpiperidin-4-amine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD07690605
CAS Number: 150514-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR