2-Chloro-6-methylnicotinic acid

Supplier: Matrix Scientific

027557-5G 027557-25G
101824-410EA 27.69 USD
101824-410 101824-412
2-Chloro-6-methylnicotinic acid
2-Chloro-6-methylnicotinic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
Melting Pt: 157-162°C
MDL Number: MFCD00134165
CAS Number: 30529-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR