1,5-Dimethyl-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

027540-1G 027540-5G
101824-378EA 78.46 USD
101824-378 101824-380
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00085040
CAS Number: 5744-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR