4-Bromo-1H-pyrazole ≥97%

Supplier: Matrix Scientific

027407-25G 027407-100G
101824-128EA 33.85 USD
101824-128 101824-130
4-Bromo-1H-pyrazole ≥97%
4-Bromo-1H-pyrazole

Hazard: Irritant

Formula: C₃H₃BrN₂
MW: 146.97 g/mol
MDL Number: MFCD00075602
CAS Number: 2075-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR