4-((1H-1,2,4-Triazol-1-yl)methyl)aniline

Supplier: Matrix Scientific

027381-5G 027381-25G
101824-068EA 21.54 USD
101824-068 101824-070
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline
4-((1H-1,2,4-Triazol-1-yl)methyl)aniline

Hazard: Irritant

Formula: C₉H₁₀N₄
MW: 174.21 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD00973306
CAS Number: 119192-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR