1-Benzyl-3-piperidone hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Benzylpiperidin-3-one hydrochloride
027372-5G
027372-25G
101824-046EA
44.62
USD
101824-046
102832-964
1-Benzyl-3-piperidone hydrochloride ≥95%
1-Benzyl-3-piperidone hydrochloride
Purity: 95%
CAS: 50606-58-1
MDL Number: MFCD00012791
Molecular Formula: C12H16ClNO
Molecular Weight: 225,72
Boiling Point: 294°C
Hazard: Toxic/Refrigerate
Formula:
C₁₂H₁₅NO·HCl MW: 225.72 g/mol Boiling Pt: 294 °C Melting Pt: 170 °C Flash Pt: 123.3 °C |
MDL Number:
MFCD00012791 CAS Number: 50606-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...