(3-Methoxyphenoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(3-Methoxyphenoxy)acetic acid

027329-5G 027329-25G
101823-960EA 73.85 USD
101823-960 101823-962
(3-Methoxyphenoxy)acetic acid
(3-Methoxyphenoxy)acetic acid

Hazard: Irritant

Formula: (C₆H₄(OCH₃))OCH₂COOH
MW: 182.18 g/mol
MDL Number: MFCD00014357
CAS Number: 2088-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR